Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031727.2 | NP_113915.2 | 3 | 389 | ||
XM_006249185.2 | XP_006249247.1 | 2 | 3481 | |||
GenBank mRNA | BF565872.1 | - | 2 | 155 | ||
CB726578.1 | - | 3 | 389 | |||
D31873.1 | - | 3 | 414 |