Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031802.1 | NP_113990.1 | 18 | 2708 | ||
GenBank mRNA | AF058795.1 | - | 18 | 2708 | ||
AF074482.1 | - | 18 | 2558 | |||
AF109405.1 | - | 18 | 2675 |
Set Membership: |
G-protein coupled receptor |