Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032462.2 | NP_115851.1 | 2, 3 | 204 | ||
GenBank mRNA | AB043892.1 | - | 1, 2 | 183 | ||
AF297118.1 | - | 2, 3 | 206 |