Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032083.1 | NP_114472.1 | 1, 2 | 664 | ||
XM_006234386.2 | XP_006234448.1 | 7, 8 | 1380 | |||
XM_006234388.2 | XP_006234450.1 | 7, 8 | 620 | |||
XM_006234389.2 | XP_006234451.1 | 7, 8 | 1323 | |||
XM_008761932.1 | XP_008760154.1 | 4, 5 | 1380 | |||
GenBank mRNA | BC094519.1 | - | 1, 2 | 713 | 7315609 | |
X67250.1 | - | 1, 2 | 664 |