Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031828.1 | NP_114016.1 | 14, 15 | 2239 | ||
GenBank mRNA | AF083341.1 | - | 14, 15 | 1675 | ||
AF135265.1 | - | 14, 15 | 2093 | |||
AY330290.1 | - | 14, 15 | 1564 | |||
AY330291.1 | - | 14, 15 | 1564 | |||
AY330292.1 | - | 14, 15 | 1573 | |||
AY330293.2 | - | 14, 15 | 1564 | |||
AY330294.1 | - | 14, 15 | 1564 | |||
AY344964.1 | - | 14, 15 | 1564 | |||
AY344965.2 | - | 13, 14 | 1976 | |||
U40603.1 | - | 14, 15 | 2239 | |||
U55995.1 | - | 14, 15 | 1717 | |||
U93052.1 | - | 14, 15 | 1665 |