Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130831.1 | NP_001124303.1 | 7 | 687 | ||
NM_003644.2 | NP_003635.2 | 3 | 436 | |||
NM_201432.1 | NP_958836.1 | 7 | 844 | |||
NM_201433.1 | NP_958839.1 | 7 | 874 | |||
XM_005256831.3 | XP_005256888.1 | 8 | 785 | |||
XM_005256832.3 | XP_005256889.1 | 7 | 655 | |||
XM_005256833.3 | XP_005256890.1 | 6 | 762 | |||
XM_005256834.3 | XP_005256891.1 | 6 | 588 | |||
XM_005256835.3 | XP_005256892.1 | 6 | 532 | |||
XM_011524044.2 | XP_011522346.1 | 7 | 679 | |||
XM_017025239.1 | XP_016880728.1 | 7 | 708 | |||
XM_017025240.1 | XP_016880729.1 | 6 | 491 | |||
XM_017025241.1 | XP_016880730.1 | 6 | 467 | |||
XM_017025242.1 | XP_016880731.1 | 6 | 641 | |||
GenBank mRNA | AB007854.1 | - | 7 | 642 | ||
AB209030.1 | - | 6 | 531 | |||
AJ224876.1 | - | 3 | 459 | |||
AK057761.1 | - | 4 | 506 | |||
AK225165.1 | - | 7 | 666 | |||
AK292987.1 | - | 7 | 660 | |||
AK293755.1 | - | 3 | 267 | |||
AK293834.1 | - | 3 | 448 | |||
AK294829.1 | - | 6 | 635 | |||
AK307987.1 | - | 8 | 696 | |||
AK309722.1 | - | 7 | 695 | |||
AK315162.1 | - | 3 | 449 | |||
AY327406.1 | - | 7 | 874 | |||
BC001152.1 | - | 7 | 667 | 3353809 | ||
BC006454.1 | - | 7 | 844 | 3547000 | ||
BT006891.1 | - | 6 | 521 |