Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053345.1 | NP_445797.1 | 2 | 166 | ||
XM_006243380.2 | XP_006243442.1 | 3 | 316 | |||
GenBank mRNA | AF000944.1 | - | 2 | 166 | ||
BC168727.1 | - | 3 | 302 | 6888477 | ||
FQ209546.1 | - | 321 | ||||
FQ212213.1 | - | 3 | 301 | |||
FQ221947.1 | - | 3 | 307 | |||
FQ228685.1 | - | 3 | 321 |