Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053437.1 | NP_445889.1 | 1 | 127 | ||
GenBank mRNA | AF296131.1 | - | 1 | 127 |