Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNOT8 SIRNA Assays ›
Gene Symbol:
CNOT8
Gene Name:
CCR4-NOT transcription complex subunit 8
Gene Aliases:
CAF1, CALIF, Caf1b, POP2, hCAF1
Chromosome Location:
Chr.5: 154857952 - 154876792 on Build GRCh38
UniGene ID:
Hs.26703
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5497
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNOT8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301073.1
NP_001288002.1
7
2009
NM_001301074.1
NP_001288003.1
6
1847
NM_001301075.1
NP_001288004.1
6
1863
NM_001301077.1
NP_001288006.1
7
1921
NM_001301080.1
NP_001288009.1
6
1820
NM_001301082.1
NP_001288011.1
6
1533
NM_001301083.1
NP_001288012.1
5
1653
NM_001301086.1
NP_001288015.1
5
1658
NM_004779.5
NP_004770.4
7
2057
XM_005268527.2
XP_005268584.1
7
2349
XM_011537706.1
XP_011536008.1
8
2031
XM_011537708.2
XP_011536010.1
6
1534
XM_017010048.1
XP_016865537.1
8
1916
XM_017010049.1
XP_016865538.1
8
2511
XM_017010050.1
XP_016865539.1
8
2496
XM_017010051.1
XP_016865540.1
8
2546
XM_017010052.1
XP_016865541.1
7
2301
XM_017010053.1
XP_016865542.1
7
2319
XM_017010054.1
XP_016865543.1
6
1840
XM_017010055.1
XP_016865544.1
6
1872
XM_017010056.1
XP_016865545.1
6
1958
XM_017010057.1
XP_016865546.1
8
2074
XM_017010058.1
XP_016865547.1
5
1662
GenBank mRNA
AF053318.1
-
7
1931
AF087844.1
-
6
1759
AF180476.1
-
6
1760
AK025358.1
-
7
1978
AL122045.1
-
7
1938
BC017366.2
-
7
1868
3506054
BX647124.1
-
5
1400
Back To Top
More Information
Panther Classification:
Molecular Function -
mRNA polyadenylation factor
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of cell proliferation
negative regulation of cell proliferation
gene silencing by miRNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
transcription factor activity, sequence-specific DNA binding
RNA binding
poly(A)-specific ribonuclease activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0