Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270802.1 | NP_001257731.1 | 9 | 1455 | ||
NM_053483.2 | NP_445935.2 | 9 | 1430 | |||
GenBank mRNA | AJ130946.1 | - | 8 | 1334 | ||
AY779026.1 | - | 8 | 1324 | |||
BC062026.1 | - | 9 | 1430 | 5621957 | ||
BC089787.1 | - | 9 | 1381 | 7326072 |
Gene Symbol: LOC100359600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729596.4 | XP_002729642.1 | 1 | 1450 |