Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053540.2 | NP_445992.1 | 2 | 222 | ||
GenBank mRNA | AB032178.1 | - | 2 | 166 |