Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053595.2 | NP_446047.1 | 5, 6 | 717 | ||
GenBank mRNA | BC087006.1 | - | 5, 6 | 717 | 7193034 | |
L40030.1 | - | 5, 6 | 452 |