Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053605.1 | NP_446057.1 | 2 | 984 | ||
XM_006255543.2 | XP_006255605.1 | 3 | 1083 | |||
XM_008772541.1 | XP_008770763.1 | 2 | 1017 | |||
XM_008772542.1 | XP_008770764.1 | 2 | 1987 | |||
GenBank mRNA | AB000215.1 | - | 2 | 984 |