Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053580.2 | NP_446032.2 | 9 | 1454 | ||
XM_006252953.2 | XP_006253015.1 | 9 | 1492 | |||
XM_006252954.1 | XP_006253016.1 | 9 | 1440 | |||
XM_008771156.1 | XP_008769378.1 | 9 | 1379 | |||
XM_008771157.1 | XP_008769379.1 | 9 | 1538 | |||
GenBank mRNA | BC074014.1 | - | 9 | 1454 | 7104661 | |
U89529.1 | - | 9 | 1351 |