Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040271.1 | NP_001035361.1 | 5, 6 | 453 | ||
NM_053592.2 | NP_446044.1 | 6, 7 | 573 | |||
GenBank mRNA | CB325379.1 | - | 5, 6 | 467 | ||
FQ220451.1 | - | 608 | ||||
FQ233848.1 | - | 1, 2 | 73 | |||
U64030.2 | - | 6, 7 | 573 |