Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053688.2 | NP_446140.1 | 1, 2 | 151 | ||
XM_006237558.2 | XP_006237620.1 | 2, 3 | 1479 | |||
GenBank mRNA | AF169390.1 | - | 1, 2 | 141 |