Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053656.2 | NP_446108.2 | 8, 9 | 913 | ||
XM_006249515.2 | XP_006249577.1 | 8, 9 | 1106 | |||
XM_006249516.2 | XP_006249578.1 | 8, 9 | 1105 | |||
XM_006249517.2 | XP_006249579.1 | 8, 9 | 1068 | |||
XM_008769305.1 | XP_008767527.1 | 7, 8 | 989 | |||
XM_008769306.1 | XP_008767528.1 | 8, 9 | 782 | |||
XM_008769307.1 | XP_008767529.1 | 6, 7 | 854 | |||
GenBank mRNA | AF013241.1 | - | 1, 2 | 138 | ||
AF020756.1 | - | 7, 8 | 813 | |||
AF020757.1 | - | 7, 8 | 859 | |||
AF020758.1 | - | 6, 7 | 953 | |||
AF020759.1 | - | 8, 9 | 875 | |||
AF064549.1 | - | 7, 8 | 950 | |||
U14414.1 | - | 8, 9 | 913 | |||
Y10474.1 | - | 8, 9 | 895 | |||
Y10475.1 | - | 8, 9 | 949 |