Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053840.2 | NP_446292.2 | 571 | |||
GenBank mRNA | BC078768.1 | - | 571 | 7133071 | ||
M33821.1 | - | 669 | ||||
M33822.1 | - | 656 | ||||
X15443.1 | - | 577 |