Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053846.1 | NP_446298.1 | 4, 5 | 1074 | ||
XM_008760057.1 | XP_008758279.1 | 5, 6 | 1328 | |||
XM_008760058.1 | XP_008758280.1 | 5, 6 | 1328 | |||
XM_008760059.1 | XP_008758281.1 | 5, 6 | 1328 | |||
XM_008760060.1 | XP_008758282.1 | 5, 6 | 1328 | |||
XM_008760061.1 | XP_008758283.1 | 5, 6 | 1328 | |||
XM_008760062.1 | XP_008758284.1 | 5, 6 | 1328 | |||
XM_008760063.1 | XP_008758285.1 | 5, 6 | 1328 | |||
GenBank mRNA | M96376.1 | - | 4, 5 | 1074 |