Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053850.1 | NP_446302.1 | 5 | 498 | ||
XM_006234894.2 | XP_006234956.1 | 6 | 638 | |||
XM_006234895.2 | XP_006234957.1 | 6 | 623 | |||
GenBank mRNA | BC078766.1 | - | 5 | 510 | 7126157 | |
FQ213399.1 | - | 6 | 577 | |||
FQ230935.1 | - | 5 | 457 | |||
FQ231949.1 | - | 6 | 579 | |||
M81681.1 | - | 5 | 498 |