Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053857.2 | NP_446309.1 | 1, 2 | 202 | ||
GenBank mRNA | U05014.1 | - | 1, 2 | 202 |