Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053948.4 | NP_446400.1 | 3 | 272 | ||
XM_006230973.1 | XP_006231035.1 | 5 | 557 | |||
GenBank mRNA | BC060550.1 | - | 3 | 228 | 6890594 | |
BC060595.1 | - | 5 | 566 | 6888231 | ||
FQ223906.1 | - | 5 | 562 | |||
FQ224086.1 | - | 3 | 305 | |||
FQ230390.1 | - | 3 | 235 | |||
FQ232354.1 | - | 3 | 276 | |||
Z71925.1 | - | 3 | 273 |