Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053979.2 | NP_446431.1 | 5 | 546 | ||
XM_008761696.1 | XP_008759918.1 | 5 | 618 | |||
GenBank mRNA | BC097294.1 | - | 5 | 546 | 7458249 | |
X78604.1 | - | 5 | 395 |