Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057129.1 | NP_476470.1 | 1, 2 | 110 | ||
GenBank mRNA | D83231.1 | - | 1, 2 | 110 |