Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277249.1 | NP_001264178.1 | 3, 4 | 315 | ||
NM_001277250.1 | NP_001264179.1 | 3, 4 | 315 | |||
NM_001277251.1 | NP_001264180.1 | 4, 5 | 374 | |||
NM_001277252.1 | NP_001264181.1 | 3, 4 | 255 | |||
NM_001277253.1 | NP_001264182.1 | 3, 4 | 367 | |||
NM_057143.2 | NP_476484.1 | 3, 4 | 497 | |||
GenBank mRNA | AF157511.1 | - | 3, 4 | 497 | ||
AF157512.1 | - | 3, 4 | 251 | |||
AJ007291.1 | - | 4, 5 | 422 | |||
BC091128.1 | - | 3, 4 | 288 | 7376914 | ||
FM055257.1 | - | 3, 4 | 285 | |||
FM061417.1 | - | 3, 4 | 236 | |||
FM069664.1 | - | 3, 4 | 306 | |||
FQ220895.1 | - | 3, 4 | 367 | |||
FQ223564.1 | - | 3, 4 | 365 | |||
FQ224480.1 | - | 4, 5 | 358 |