Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256864.1 | NP_001243793.1 | 2 | 481 | ||
NM_001256865.1 | NP_001243794.1 | 3 | 429 | |||
NM_014787.3 | NP_055602.1 | 2 | 316 | |||
GenBank mRNA | AB007942.1 | - | 2 | 256 | ||
AK296408.1 | - | 3 | 383 | |||
BC051722.1 | - | 2 | 265 | |||
BC109279.2 | - | 2 | 414 | 40028812 | ||
BC109280.1 | - | 2 | 417 | 40028814 | ||
BP311649.1 | - | 2 | 282 | |||
BX647203.1 | - | 2 | 470 | |||
DC332732.1 | - | 3 | 429 | |||
DR002818.1 | - | 2 | 489 |