Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270726.1 | NP_001257655.1 | 4 | 592 | ||
NM_001270727.1 | NP_001257656.1 | 4 | 592 | |||
NM_001270729.1 | NP_001257658.1 | 4 | 606 | |||
NM_001270730.1 | NP_001257659.1 | 3 | 435 | |||
NM_015367.3 | NP_056182.2 | 5 | 763 | |||
XM_011546119.1 | XP_011544421.1 | 5 | 1126 | |||
XM_011546120.1 | XP_011544422.1 | 5 | 1128 | |||
XM_011546121.2 | XP_011544423.1 | 6 | 1111 | |||
XM_017028725.1 | XP_016884214.1 | 5 | 1126 | |||
XM_017028726.1 | XP_016884215.1 | 7 | 1412 | |||
XM_017028727.1 | XP_016884216.1 | 6 | 705 | |||
XM_017028728.1 | XP_016884217.1 | 5 | 1140 | |||
XM_017028731.1 | XP_016884220.1 | 4 | 969 | |||
XM_017028733.1 | XP_016884222.1 | 5 | 763 | |||
GenBank mRNA | AF146568.1 | - | 2 | 184 | ||
AF246665.1 | - | 6 | 1149 | |||
AF325209.1 | - | 4 | 410 | |||
AK056248.1 | - | 4 | 449 | |||
AK314489.1 | - | 5 | 590 | |||
BC007658.2 | - | 5 | 522 | 3343994 | ||
BX354578.2 | - | 4 | 471 | |||
BX407004.2 | - | 4 | 345 | |||
CR456521.1 | - | 5 | 666 | |||
DA154984.1 | - | 3 | 262 | |||
DA478149.1 | - | 4 | 422 |