Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078622.2 | NP_511177.2 | 3 | 293 | ||
XM_006248444.2 | XP_006248506.1 | 3 | 446 | |||
XM_008768694.1 | XP_008766916.1 | 3 | 644 | |||
GenBank mRNA | AW919077.1 | - | 3 | 294 | ||
BC085713.1 | - | 3 | 291 | 7127059 | ||
L13245.1 | - | 2 | 190 | |||
M36071.1 | - | 3 | 290 | |||
U03490.1 | - | 3 | 255 |