Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057194.1 | NP_476542.1 | 5, 6 | 789 | ||
XM_006243536.1 | XP_006243598.1 | 6, 7 | 864 | |||
XM_006243537.1 | XP_006243599.1 | 6, 7 | 859 | |||
XM_006243538.2 | XP_006243600.1 | 5, 6 | 811 | |||
GenBank mRNA | AY024347.1 | - | 5, 6 | 789 | ||
BC085715.1 | - | 6, 7 | 788 | 7109899 |
Gene Symbol: Plscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014094.1 | NP_001014116.1 | 5, 6 | 650 | ||
XM_008766450.1 | XP_008764672.1 | 5, 6 | 897 | |||
GenBank mRNA | BC079090.1 | - | 5, 6 | 650 | 7123519 |