Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014369.3 | NP_055184.2 | 3 | 352 | ||
XM_006712417.1 | XP_006712480.1 | 3 | 347 | |||
GenBank mRNA | DN995571.1 | - | 3 | 360 | ||
X79568.1 | - | 3 | 294 |