Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014372.4 | NP_055187.1 | 2 | 687 | ||
GenBank mRNA | AB024703.1 | - | 2 | 330 | ||
AF151881.1 | - | 2 | 330 | |||
AK293047.1 | - | 4 | 516 | |||
AK313140.1 | - | 2 | 449 | |||
BC020964.2 | - | 2 | 282 | 3917224 | ||
BC047654.1 | - | 2 | 655 | 5261966 | ||
DB265274.1 | - | 4 | 517 |