Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040101.1 | NP_001035190.1 | 3, 4 | 661 | ||
NM_001287763.1 | NP_001274692.1 | 4, 5 | 716 | |||
NM_001287764.1 | NP_001274693.1 | 3, 4 | 607 | |||
NM_014392.4 | NP_055207.1 | 3, 4 | 484 | |||
XM_017008022.1 | XP_016863511.1 | 6, 7 | 1901 | |||
GenBank mRNA | AK094573.1 | - | 8, 9 | 2324 | ||
AK125172.1 | - | 4, 5 | 698 | |||
AK312288.1 | - | 3, 4 | 343 | |||
AX775767.1 | - | 3, 4 | 408 | |||
BC001745.1 | - | 3, 4 | 408 | 3510777 | ||
BC034227.1 | - | 6, 7 | 1357 | 5265974 | ||
DA746130.1 | - | 3, 4 | 312 | |||
DA773692.1 | - | 3, 4 | 480 |