Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133296.1 | NP_579830.1 | 4 | 537 | ||
GenBank mRNA | FQ211786.1 | - | 645 | |||
S76742.1 | - | 4 | 537 |