Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133522.1 | NP_598206.1 | 3 | 1740 | ||
XM_006241966.2 | XP_006242028.1 | 2 | 1942 | |||
GenBank mRNA | X63574.1 | - | 3 | 1740 |
Gene Symbol: Ttbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271230.1 | NP_001258159.1 | 13 | 2225 |
Set Membership: |
G-protein coupled receptor |