Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlxipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133552.1 | NP_598236.1 | 2 | 372 | ||
GenBank mRNA | AB074517.1 | - | 2 | 372 | ||
FN432819.1 | - | 2 | 339 |