Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321708.1 | NP_001308637.1 | 1 | 367 | ||
NM_001321710.1 | NP_001308639.1 | 1 | 367 | |||
NM_004717.3 | NP_004708.1 | 1 | 367 | |||
XM_011516682.2 | XP_011514984.2 | 1 | 364 | |||
XM_017012780.1 | XP_016868269.1 | 1 | 365 | |||
XM_017012781.1 | XP_016868270.1 | 1 | 364 | |||
XM_017012782.1 | XP_016868271.1 | 1 | 364 | |||
XM_017012783.1 | XP_016868272.1 | 1 | 364 | |||
XM_017012784.1 | XP_016868273.1 | 1 | 364 | |||
XM_017012787.1 | XP_016868276.1 | 1 | 26 | |||
XM_017012788.1 | XP_016868277.1 | 1 | 23 | |||
GenBank mRNA | AB209167.1 | - | 1 | 39 | ||
DR002813.1 | - | 1 | 372 |