Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5722
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
5
751
NM_001270728.1
NP_001257657.1
3
587
NM_001270729.1
NP_001257658.1
5
765
NM_001270730.1
NP_001257659.1
4
594
NM_001270731.1
NP_001257660.1
5
852
NM_001270732.1
NP_001257661.1
4
695
NM_015367.3
NP_056182.2
6
922
XM_011546119.1
XP_011544421.1
6
1285
XM_011546120.1
XP_011544422.1
6
1287
XM_011546121.2
XP_011544423.1
7
1270
XM_017028726.1
XP_016884215.1
8
1571
XM_017028728.1
XP_016884217.1
6
1299
XM_017028729.1
XP_016884218.1
6
794
XM_017028730.1
XP_016884219.1
4
508
XM_017028731.1
XP_016884220.1
5
1128
XM_017028732.1
XP_016884221.1
3
371
XM_017028733.1
XP_016884222.1
6
922
XM_017028734.1
XP_016884223.1
5
1229
GenBank mRNA
AF146568.1
-
3
343
AF246665.1
-
7
1308
AF325209.1
-
5
569
AK056248.1
-
5
608
AK291449.1
-
4
515
AK294286.1
-
3
414
AK314489.1
-
6
749
AL133029.1
-
1
24
BC003032.1
-
2
182
BC007658.2
-
6
681
3343994
BE900458.1
-
5
636
BI560132.1
-
5
708
BX407004.2
-
5
504
CR456521.1
-
6
825
DA154984.1
-
4
421
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0