Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5727
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
3
345
NM_001042358.4
NP_001035817.1
2
203
NM_001042359.3
NP_001035818.1
2
203
NM_001042361.4
NP_001035820.1
2
203
NM_001042362.3
NP_001035821.1
2
203
NM_001042363.4
NP_001035822.1
2
188
NM_001320683.1
NP_001307612.1
3
360
NM_001320684.1
NP_001307613.1
3
360
NM_001320689.1
NP_001307618.1
2
244
NM_015605.8
NP_056420.3
3
345
XM_011539605.2
XP_011537907.1
3
489
XM_011539606.2
XP_011537908.1
3
1581
XM_011539607.2
XP_011537909.1
3
480
XM_017016042.1
XP_016871531.1
3
324
XM_017016043.1
XP_016871532.1
3
324
XM_017016046.1
XP_016871535.1
3
324
XM_017016047.1
XP_016871536.1
3
324
XM_017016048.1
XP_016871537.1
3
324
XM_017016051.1
XP_016871540.1
3
324
XM_017016052.1
XP_016871541.1
3
324
XM_017016053.1
XP_016871542.1
3
324
XM_017016055.1
XP_016871544.1
3
324
XM_017016056.1
XP_016871545.1
2
332
XM_017016060.1
XP_016871549.1
3
324
XM_017016063.1
XP_016871552.1
3
365
XM_017016065.1
XP_016871554.1
2
167
GenBank mRNA
AY704141.1
-
2
144
AY704142.1
-
2
144
AY704144.1
-
2
144
AY704145.1
-
2
144
AY704146.1
-
2
193
AY704147.1
-
3
301
AY704148.1
-
3
301
AY704149.1
-
2
193
AY704155.1
-
2
234
AY704156.1
-
2
185
AY753191.1
-
2
203
BC093750.1
-
2
199
7939595
BC113473.1
-
2
199
8322525
BC143394.1
-
2
199
BC171725.1
-
2
199
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0