Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133569.1 | NP_598253.1 | 4 | 1476 | ||
GenBank mRNA | AF159049.1 | - | 4 | 1476 |