Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321708.1 | NP_001308637.1 | 33 | 4464 | ||
NM_001321709.1 | NP_001308638.1 | 34 | 4242 | |||
NM_001321710.1 | NP_001308639.1 | 33 | 4361 | |||
NM_004717.3 | NP_004708.1 | 34 | 4488 | |||
XM_011516684.2 | XP_011514986.2 | 27 | 11708 | |||
XM_017012780.1 | XP_016868269.1 | 34 | 4525 | |||
XM_017012781.1 | XP_016868270.1 | 34 | 4509 | |||
XM_017012782.1 | XP_016868271.1 | 33 | 4410 | |||
XM_017012783.1 | XP_016868272.1 | 33 | 4401 | |||
XM_017012784.1 | XP_016868273.1 | 32 | 4335 | |||
XM_017012785.1 | XP_016868274.1 | 32 | 4712 | |||
XM_017012786.1 | XP_016868275.1 | 34 | 4212 | |||
XM_017012787.1 | XP_016868276.1 | 33 | 4077 | |||
XM_017012788.1 | XP_016868277.1 | 34 | 4195 | |||
XM_017012789.1 | XP_016868278.1 | 20 | 2921 | |||
GenBank mRNA | AB209167.1 | - | 33 | 4033 | ||
AK091081.1 | - | 7 | 2106 |