Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134387.1 | NP_599214.1 | 6 | 505 | ||
XM_008768465.1 | XP_008766687.1 | 6 | 551 | |||
XM_008768466.1 | XP_008766688.1 | 6 | 551 | |||
XM_008768467.1 | XP_008766689.1 | 6 | 551 | |||
GenBank mRNA | AB061719.1 | - | 6 | 505 | ||
FQ233401.1 | - | 6 | 504 |