Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134467.1 | NP_604462.1 | 5 | 564 | ||
XM_006238012.2 | XP_006238074.1 | 5 | 814 | |||
XM_006238014.2 | XP_006238076.1 | 5 | 712 | |||
XM_006238015.2 | XP_006238077.1 | 4 | 664 | |||
XM_006238016.2 | XP_006238078.1 | 4 | 911 | |||
XM_006238017.2 | XP_006238079.1 | 4 | 1684 | |||
XM_006238018.2 | XP_006238080.1 | 5 | 752 | |||
XM_006238020.2 | XP_006238082.1 | 5 | 557 | |||
XM_008763664.1 | XP_008761886.1 | 4 | 1338 | |||
XM_008763665.1 | XP_008761887.1 | 4 | 1251 | |||
XM_008763666.1 | XP_008761888.1 | 4 | 1236 | |||
XM_008763667.1 | XP_008761889.1 | 6 | 707 | |||
XM_008763668.1 | XP_008761890.1 | 5 | 650 | |||
GenBank mRNA | AF480444.1 | - | 5 | 564 | ||
BC083548.1 | - | 4 | 479 | 7134112 |