Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138510.1 | NP_612519.1 | 3 | 364 | ||
GenBank mRNA | BC086579.1 | - | 3 | 363 | ||
BC100248.1 | - | 3 | 358 | 7307214 | ||
D14424.1 | - | 3 | 349 | |||
L32601.1 | - | 3 | 364 |