Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 5 | 901 | ||
GenBank mRNA | AJ298278.1 | - | 5 | 901 | ||
BC083176.1 | - | 5 | 1145 | 7097075 | ||
FQ225555.1 | - | 2 | 189 | |||
FQ231126.1 | - | 2 | 189 | |||
FQ231168.1 | - | 2 | 186 | |||
FQ233364.1 | - | 2 | 189 | |||
FQ233754.1 | - | 1 | 30 | |||
FQ234731.1 | - | 2 | 118 | |||
FQ235339.1 | - | 2 | 178 |