Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016531.5 | NP_057615.3 | 3 | 853 | ||
XM_017008279.1 | XP_016863768.1 | 4 | 1047 | |||
GenBank mRNA | AB024523.1 | - | 3 | 576 | ||
AF285837.1 | - | 3 | 680 | |||
BC030662.1 | - | 3 | 828 | 5262512 | ||
BC051687.1 | - | 3 | 825 | 5303520 |