Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138532.1 | NP_612541.1 | 13 | 1338 | ||
XM_008769600.1 | XP_008767822.1 | 11 | 1301 | |||
GenBank mRNA | AF202049.1 | - | 13 | 1338 | ||
BC060314.1 | - | 13 | 1359 | 6918381 |