Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138613.1 | NP_619516.1 | 3 | 289 | ||
XM_006246441.2 | XP_006246503.1 | 5 | 1401 | |||
XM_006246445.2 | XP_006246507.1 | 3 | 292 | |||
XM_008767774.1 | XP_008765996.1 | 5 | 1829 | |||
XM_008767775.1 | XP_008765997.1 | 5 | 1829 | |||
XM_008767776.1 | XP_008765998.1 | 5 | 1538 | |||
XM_008767777.1 | XP_008765999.1 | 4 | 474 | |||
GenBank mRNA | AF054586.1 | - | 3 | 289 | ||
FQ212014.1 | - | 2 | 205 |