Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138880.2 | NP_620235.1 | 1 | 87 | ||
GenBank mRNA | AF010466.1 | - | 1 | 87 |