Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138547.2 | NP_612556.1 | 2 | 143 | ||
GenBank mRNA | BC091123.1 | - | 2 | 143 | 7372929 | |
D17310.1 | - | 2 | 331 | |||
M64393.1 | - | 2 | 142 | |||
S57790.1 | - | 2 | 101 |