Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138862.3 | NP_620217.2 | 10, 11 | 1207 | ||
XM_006240413.2 | XP_006240475.1 | 9, 10 | 1190 | |||
XM_006240414.2 | XP_006240476.1 | 10, 11 | 1282 | |||
XM_006240415.2 | XP_006240477.1 | 8, 9 | 1115 | |||
XM_008764737.1 | XP_008762959.1 | 9, 10 | 1161 | |||
GenBank mRNA | BC078728.1 | - | 10, 11 | 933 | 7114267 |